Corning™ Axygen™ 96-well PCR Microplates

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.

Manufacturer: Corning™ PCR96FSO

 View more versions of this product

Catalog No. 14-222-329

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Equipment) Standard 96 well blocks
    Type Flat top
    Certifications/Compliance RNase-, DNase-free
    Well Design Round
    Material 99.9% pure virgin polypropylene,Polypropylene
    Color Orange, clear well
    Barcode No
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Skirt Style Full
    Sterile No,No
    Volume (Metric) Well 200μL
    Product Type 96-Well PCR Microplate
    Applications PCR thermal transfer, RT-PCR applications
    Well Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit