Axygen™ 96-well Full-Skirted PCR Microplates Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Axygen Scientific PCR96FSV

Catalog No. 14-222-331

  Check Availability
Add to Cart

    Specifications

    Barcode No
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Material Polypropylene,Polypropylene
    Skirt Style Full
    Sterile No,No
    Certifications/Compliance RNase, DNase, pyrogen-free
    Volume (Metric) Well 200μL
    Well Color Clear
    Color Violet
    Product Type 96-Well PCR Microplate
    Applications PCR thermal transfer, RT-PCR applications

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit