Axygen™ 96-Well Half-Skirt PCR Microplates

Half-skirt amplification plate with single notch

Manufacturer: Axygen Scientific PCR96M2HSB

 View more versions of this product

Catalog No. 14-223-334

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Material Polypropylene,Polypropylene
    Skirt Style Half
    Sterile No,No
    Certifications/Compliance RNase, DNase-free
    Volume (Metric) Well 300μL
    Well Color Clear
    Well Design Round
    Color Blue
    Product Type 96-Well PCR Microplate
    Applications PCR thermal transfer, RT-PCR applications
    Well Shape Round

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit