Corning™ Axygen™ 96-Well 100µL PCR Microplates Available on GSA/VA Contract for Federal Government customers only.

Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Corning™ Axygen™ 96-well 100µL PCR Microplates are specifically designed to be compatible with Roche Light Cycler 480.

Manufacturer: Corning™ PCR96LC480WBC

 View more versions of this product

Catalog No. 14-222-336

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Roche Light Cycler 480
    Includes Ultra Clear sealing film, bar codes, printed alphanumeric grid
    Material 99.9% pure virgin polypropylene,Polypropylene with sealing film
    Skirt Style Half skirted
    Certifications/Compliance RNase-, DNase-free
    Barcode Yes
    For Use With (Application) PCR thermal transfer, RT-PCR applications
    Sterile No,No
    Volume (Metric) Well 100μL
    Well Design Round
    Color White
    Product Type 96-Well PCR Microplate
    Applications PCR thermal transfer, RT-PCR applications
    Well Shape Round
    Well Color White

    • Nonpyrogenic
    • Certified RNase- and DNase-free
    • Nonsterile
    • Large print alphanumeric grid for ease of well identification




     

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit