Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Axygen™ 96-well PCR Microplates
Constructed with ultra-thin and consistent wall thickness to allow precise thermal transfer for optimum results. Axygen™ 96-well PCR Microplates are the ideal option for use in PCR thermal transfer and Real-Time PCR applications.
$155.00 - $514.00
Specifications
Certifications/Compliance | RNase-, DNase-free |
---|---|
Format | Standard |
Array | 8 x 12 |
Color | Clear |
Material | Polypropylene |
Catalog Number | Mfr. No. | Skirt Style | For Use With (Equipment) | Sterility | Barcode | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Skirt Style | For Use With (Equipment) | Sterility | Barcode | Price | Quantity & Availability | ||
14-224-002
|
Axygen™
PCR96M2HSCPBC |
Half | ABI PCR 2700, 2720, 9700, 9800, 7300, 7500, 7900 and ABI sequencing units | Non-sterile |
Case of 50 for $514.00
|
|
14-224-001
|
Axygen™
PCR96ABCPBC |
Elevated, Semi | ABI Real time PCR 7300, 7500, 7900, qPCR and ABI Sequencing | Non-sterile | Includes Barcode |
Case of 50 for $631.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
14-222-327
|
Axygen™
PCR96FSCS |
Full | Standard 96 well blocks | Sterile |
Pack of 10 for $155.00
Case of 5 PK for $691.00 |
![]() |
14-224-003
|
Axygen™
PCR96FSCPBC |
Full | Standard 96 well blocks | Non-sterile | Includes Barcode |
Case of 50 for $514.00
|
|
Description
Axygen™ 96 well elevated skirt bar coded PCR microplate is designed to fit ABI™ Real Time PCR thermal cyclers and sequencers. Ultra-thin and consistent wall thickness allows precise thermal transfer for optimum results. Can be sealed with 8/12 strip caps, Axymat or sealing film. Bar code on the long side of the plate between well H1-H12.Specifications
RNase-, DNase-free | |
8 x 12 | |
Polypropylene | |
Round |
Standard | |
Clear | |
96 | |
PCR thermal transfer, RT-PCR applications |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title