Axygen™ Aerosol Filter Tips, Non-sterile
Prevent cross-contamination of samples. Axygen™ Aerosol Filter Tips, Non-sterile feature a universal fit. All tips comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Compatible with a variety of pipettes and available in a range of filter sizes.
Manufacturer: Axygen™ TF350
 View more versions of this product
Catalog No. 14-222-786
  Check Availability
Add to Cart
    Specifications
    Product Type Aerosol Barrier Tip
    Material Polypropylene
    Packaging Bulk: 1000 Tips/Pk/, 10 Packs/Cs.
    Sterility Nonsterile
    Certifications/Compliance DNase, RNase, pyrogen free
    USP 3127 (Heavy Metal Contamination).
    Tip Style Barrier, Universal Fit
    Volume (Metric) 1 to 300μL
    Color Clear
    Technologically advanced filter barrier pipette tips
    • Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
    • Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
    • Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
    • Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
    • Packaged in sealed-bottom racks with lids to prevent contamination
  • Description & Specifications
Related Products
Product Content Feedback