Axygen™ 10µL Microvolume Filter Tips Available on GSA/VA Contract for Federal Government customers only.

Prevent cross-contamination with robotic filter barrier tips. Axygen™ 10µL Microvolume Filter Tips are available with Maxymum Recovery&trade technology to allow recovery of costly samples with improved precision and accuracy. Extra long and sterilized pipette tips are also available.

Manufacturer: Axygen™ TF400RS

 View more versions of this product

Catalog No. 14-222-793

  Check Availability
Add to Cart

    Specifications

    Product Type Microvolume Tip
    Material Polypropylene
    For Use With (Equipment) Eppendorf™ Research™ 0.5 to 10μL
    Sterility Sterile
    Tip Style Eppendorf style, Aerosol Barrier
    Packaging Rack
    Quantity 4800 Case
    Certifications/Compliance RNase, DNase-free
    Comply with USP 3127 (Heavy Metal Contamination)
    For Use With (Application) Allows recovery of costly samples with improved precision and accuracy
    Volume (Metric) 0.5 to 10μL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit