Learn More
Axygen™ Aerosol Filter Tips, Sterile Available on GSA/VA Contract for Federal Government customers only.
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation, comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Available in a wide variety of tip styles.
Manufacturer:  Axygen™ TF50LRS
 View more versions of this product
Catalog No. 14-222-799
  Check Availability
Add to cart
Specifications
Description
Technologically advanced filter barrier pipette tips
  • Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
  • Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
  • Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
  • Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
  • Packaged in sealed-bottom racks with lids to prevent contamination
  • Wide variety of tip styles available in a range of packaging quantities:
    • Maxymum Recovery™ technology provides excellent accuracy and prevents wastage of precious samples
    • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
    • Other options include: universal fit, matrix and Eppendorf style
Description & Specifications
Specifications
Aerosol Barrier Tip
Sterile
Barrier; Maxymum Recovery
Racked: 96 Tips/rack, 10 Racks/Pk., 5 Packs/Cs.
DNase, RNase, pyrogen free
USP 3127 (Heavy Metal Contamination)
0.5 to 50μL
Clear
Polypropylene
4,800 Tips/Case
Finnpipette™ 5 to 40μL