Axygen™ Aerosol Filter Tips, Sterile Available on GSA/VA Contract for Federal Government customers only.

Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation, comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Available in a wide variety of tip styles.

Manufacturer: Axygen™ TF100RS

 View more versions of this product

Catalog No. 14-222-770

  Check Availability
Add to Cart

    Specifications

    Product Type Aerosol Barrier Tip
    Material Polypropylene
    Quantity 4,800 Tips/Case
    For Use With (Equipment) Pipetman™ P-100
    Sterility Sterile
    Packaging Racked: 96 Tips/rack, 10 Racks/Pk., 5 Packs/Cs.
    Certifications/Compliance DNase, RNase, pyrogen free
    USP 3127 (Heavy Metal Contamination)
    Tip Style Barrier, Universal Fit
    Volume (Metric) 1 to 100μL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit