Axygen™ Aerosol Filter Tips, Sterile Available on GSA/VA Contract for Federal Government customers only.
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation, comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Available in a wide variety of tip styles.
Manufacturer: Axygen™ TF1200CLRS
 View more versions of this product
Catalog No. 14-222-771
  Check Availability
Add to Cart
    Specifications
    Product Type Aerosol Barrier Tip
    Material Polypropylene
    Quantity 3,456 Tips/Case
    Sterility Sterile
    Certifications/Compliance DNase, RNase, pyrogen free
    USP 3127 (Heavy Metal Contamination)
    Tip Style Barrier; Maxymum Recovery
    Packaging Racked: 96 Tips/rack, 12 Racks/Pk., 3 Packs/Cs.
    Volume (Metric) 100 to 1100μL
    Color Clear
  • Description & Specifications
Related Products
Product Content Feedback