Axygen™ Agarose LE Available on GSA/VA Contract for Federal Government customers only.

Axygen™ Agarose LE is a high quality molecular biology grade agarose suitable for analytical and preparative electrophoresis of nucleic acids.

Manufacturer: Axygen Scientific AGRLE500

 View more versions of this product

Catalog No. 14-223-080

  Check Availability
Add to Cart

    Specifications

    Dnase Free Yes
    Material Agarose
    RNase Free Yes
    For Use With (Application) Electrophoresis
    Melting Temperature 88°C
    Quantity 500g
    Storage Requirements 15° to 25°C

    • Nucleic acid separation with Agarose LE is between 0.2 to 15kbp depending on the concentration of Agarose LE
    • Nucleic acid fragments separated with Agarose LE can be blotted to nylon or nitrocellulose membranes
    • LE is tested for preparative electrophoresis and isolation of DNA fragments
    • Detection with non-radioactive probes, e.g. digoxigenin (DIG) – labeled nucleic acids, does not interfere with the use of Agarose LE
    DNA electrophoresis, Analysis of PCR products, Separation of restriction endonuclease digests of DNA, Electrophoresis of RNA, Protein Electrophoresis such as radial diffusion

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit