Learn More
Axygen™ Agarose LM
Axygen™ Low melting agarose allows for the recovery of undamaged nucleic acids below their denaturation temperature.
Manufacturer:  Axygen Scientific AGR-LM-50
 View more versions of this product
Catalog No. 14-223-082
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Tissue and cell culture and viral plaque assays
50g
Agarose
Yes
≥76°C
15° to 25°C
Yes
Promotions
Promotions
  • Thermo Scientific
    Maximize your separation capabilities and your budget with three ways to save on Thermo Scientific™ centrifuges. Explore the offers below to find the right solution for your lab.