Axygen™ Agarose LM Available on GSA/VA Contract for Federal Government customers only.

Axygen™ Low melting agarose allows for the recovery of undamaged nucleic acids below their denaturation temperature.

Manufacturer: Axygen Scientific AGRLM1001

 View more versions of this product

Catalog No. 14-223-043

  Check Availability
Add to Cart

    Specifications

    Quantity 10 × 100g
    Material Agarose,Agarose
    RNase Free Yes,Yes
    Dnase Free Yes
    Melting Temperature ≥76°C,≥76°C
    Storage Requirements 15° to 25°C
    For Use With (Application) Tissue and cell culture and viral plaque assays,Tissue and cell culture and viral plaque assays
    DNase Free Yes

    The low gelling temperature ensures that the agarose will be in a liquid state at a temperature range where In-Gel manipulations can be performed without prior extraction of the DNA from the gel slice. Low Melting (LM) Agarose is derivatized by organic synthesis which generates methoxylate groups from the basic agarose structure. Agarose LM is also optimized for preparative electrophoresis and isolation of DNA fragments.

    • Highest gelling/melting temperatures and gel strength
    • Electrophoresis of DNA fragments ≥1000bp
    • Suitable for tissue and cell culture and viral plaque assays
    • Ideal for preparative electrophoresis

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit