Learn More
Axygen™ Agarose MS
Axygen™ Agarose MS Molecular screening agarose for improved resolution of DNA fragments with 500bp or less, especially primer-sized fragments.
Supplier: Axygen™ AGRMS1001
Description
- Sharp finely-resolved bands
- High resolution of short PCR products and DNA fragments for improved clarity of the gel, enhancing visibility
- Stronger gel structure and higher gel strength for better handling
- The chances of gels breaking or cracking are greatly minimized, even at lower concentrations
- High gel strength allows use in blotting
- Low DNA binding for easy recovery
- At a 3% concentration, Agarose MS gives a resolution of DNA fragments similar to gels made with polyacrylamide at concentrations of 8%
Specifications
MS Agarose Gel | |
Improved resolution of DNA fragments with 500bp or less, especially primer sized fragment | |
≥65.5°C | |
Yes |
Yes | |
Agarose | |
10 x 100 g | |
15° to 25°C |