Learn More
Axygen™ Automation Tips for Agilent™/Velocity11 VPrep™ and Bravo: 384 Tips/Rack
Comprehensive line of tips for Agilent/Velocity11 automation systems
Manufacturer:  Axygen Scientific VT38410LR
 View more versions of this product
Catalog No. 14-223-525
Add to cart
Description
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Available nonsterile or sterile and filtered or non-filtered
  • Also available in Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss
Order Info
Packaging:10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
Agilent™/Velocity VPrep™ 16ST, Agilent™/Velocity VPrep™ 96ST, Agilent™/Velocity VPrep™ 384ST, Agilent™ Bravo™ 384ST
Automation
Non-sterile
Yes
PP (Polypropylene)
Maxymum Recovery™ technology for ultra-low retention
Rack
10μL
Clear
Round
DNase, RNase, and Pyrogen free
Agilent™/Velocity VPrep™ 16ST, Agilent™/Velocity VPrep™ 96ST, Agilent™/Velocity VPrep™ 384ST, Agilent™ Bravo™ 384ST