Learn More
Axygen™ Automation Tips for Agilent™/Velocity11 VPrep™ and Bravo: 96 Tips/Rack
Comprehensive line of tips for Agilent/Velocity11 automation systems
$444.50
Specifications
Autoclavable Autoclavable
Material PP (Polypropylene)
Format Rack
View More Specs
Products 1
Catalog Number Mfr. No. For Use With (Equipment) Sterility Volume (Metric) Shape Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Sterility Volume (Metric) Shape Price Quantity & Availability  
14-223-511
View Documents
Axygen Scientific
VT50R
Agilent™/Velocity VPrep™ 8LT, Agilent™/Velocity VPrep™ 96LT, Agilent™ Bravo™ 96LT Non-sterile 50 μL Round
Case of 4800 for $444.50
Add to cart
Description
Description
Tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality.
  • Clear polypropylene
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Available nonsterile or sterile and filtered or non-filtered
Order Info
Packaging:10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
Autoclavable
PP (Polypropylene)
Rack