Axygen™ AxyMats™ Pierceable Sealing Mats for 24 Well Deep Well Plates

Minimize evaporation in wide range of temperatures in PCR and storage applications

Manufacturer: Axygen Scientific AM24SQ

 View more versions of this product

Catalog No. 14-222-008

  Check Availability
Add to Cart

    Specifications

    For Use With 24 well deep well plates with square wells
    No. per Pack 10.0
    Autoclavable Yes
    No. per Case 5/Pk.
    Format Sealing Mat
    Material Silicone
    Sterile No
    No. of Wells 24
    Product Type Sealing Mat

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit