Axygen™ AxyMats™ Sealing Mats for 2mL 96 Well Assay Plates with Round Wells

Minimize evaporation in wide range of temperatures in PCR and storage applications

Manufacturer: Axygen Scientific AM2MLRDS

 View more versions of this product

Catalog No. 14-222-011

  Check Availability
Add to Cart

    Specifications

    For Use With 2mL 96-well microplates
    No. per Case 10/Pk.
    Material Silicone
    Sterile Yes
    No. of Wells 96
    Autoclavable Yes
    No. per Pack 10.0
    Format Sealing Mat
    Color Natural
    Product Type Sealing Mat

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit