Axygen™ Axypet-16 Pipette Tips Available on GSA/VA Contract for Federal Government customers only.

For 1536-well pipetting

Manufacturer: Axygen Scientific T1536LR

 View more versions of this product

Catalog No. 14-223-372

  Check Availability
Add to Cart

    Specifications

    Product Type Tip
    Material Polypropylene
    Sterility Nonsterile
    Tip Style Maxymum Recovery™
    Packaging Rack
    Quantity 50 Case
    Certifications/Compliance RNase, DNase-free
    For Use With (Application) For 1536-well pipetting
    Volume (Metric) 0.5 to 30μL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit