Axygen™ Axypet-16 Pipetters Available on GSA/VA Contract for Federal Government customers only.
16-channel pipetter for 384/1536-well formats
Manufacturer: Axygen Scientific AP16
 View more versions of this product
Catalog No. 14-222-927
  Check Availability
Add to Cart
    Specifications
    Accuracy ±2.5 to 1%
    Autoclavable Yes
    Color White,White
    Number of Channels 16
    Volume (Metric) 0.5 to 10μL
  • Description & Specifications
Related Products
Product Content Feedback