Learn More
Axygen™ AxyPrep™ MAG DNA Concentrate Kit
High-quality Plant Genomic DNA
Manufacturer:  Axygen Scientific MAGCONDNA10
 View more versions of this product
Catalog No. 14-223-421
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Utilizes paramagnetic beads based procedure to purify and concentrate DNA
MAG-C1: 110μL; BB-C1: 1.5mL; Eluction Buffer: 0.5mL; Protocol Manual: 1
Concentrate Kit
10 Preps