Axygen™ AxyPrep Mag™ FFPE (DNA-RNA-miRNA) Kits Available on GSA/VA Contract for Federal Government customers only.

Isolation of nucleic acids from formalin-fixed, paraffin-embedded tissue

Manufacturer: Axygen Scientific MAGFFPEM

 View more versions of this product

Catalog No. 14-223-173

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Isolation of Nucleic Acids from 10mg of formalin-fixed, Paraffin-embedded tissue
    Format 96-Well, Tube
    Size Small (96 preps)
    Product Type Kit
    Quantity 96 Preps,96 Preps
    Product Name Kit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit