Axygen™ AxyPrep Mag™ FragmentSelect Kits

Next generation sequencing sample preparation simplified

Manufacturer: Axygen Scientific MAG-FRAG-R-1

 View all options for this product

Catalog No. 14-223-229


This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    For Use With (Application) Genomic DNA purification
    Format 96- and 384-well plate
    Includes AxyPrep™ Mag™ FragmentSelect-R paramagnetic bead solution
    No. of Reactions 15 to 30 Preps
    Product Type Kit
    Quantity 1mL

    Post DNA shearing, the library construction process for next generation sequencing mostly requires fragment selection regardless of the platform. Obtaining high recovery post fragment selection is becoming an important contributor for the reduction of sequencing bias. The AxyPrep FragmentSelect-I kit is optimized for Illumina next generation sequencing platforms and Life Technologies (SOLiD) fragment size selection needs, simplifying the library construction workflows. Where, the AxyPrep FragmentSelect-R is designed for fragment size selection needs in the Roche/454s DNA Genome sequencer library construction workflow. The AxyPrep Mag FragmentSelect kits utilize a unique paramagnetic bead technology for quick high-throughput optimized DNA size selection suitable for various next generation sequencing platforms.

    • Optimized fragment selection chemistries for Illumina, SOLiD and Roche/454
    • Versatile and flexible sequential fragment size selection: no gels needed
    • Efficient removal of primer-dimers; no need for further clean-up steps
    • Manual and automation friendly
    • Scalable: Tube, 96- and 384-well microplate formats
    • No centrifugation or filtration required
    AxyPrep FragmentSelect kits consists of two major stages:
    • 1. Optimization of AxyPrep FragmentSelect beads ratio to input DNA for desired target DNA fragment size selection
    • 2. The optimal AxyPrep FragmentSelect beads ratio is utilized for the isolation of the desired target DNA fragment size
    Note: AxyPrep FragmentSelect kits can be a one-stage process when large size DNA fragments are not a concern.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit