Axygen™ AxyPrep Mag™ PCR Clean-up Kits Available on GSA/VA Contract for Federal Government customers only.

Post PCR clean-up simplified

Manufacturer: Axygen Scientific MAGPCRCL250

 View more versions of this product

Catalog No. 14-223-153

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Efficient removal of unincorporated dNTPs, salts and Enzymes
    Format 96 and 384-well plate
    Product Type Kit
    Quantity 250mL,250mL
    Product Name Kit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit