Axygen™ AxyPrep™ Mag PCR Normalizer Kits Available on GSA/VA Contract for Federal Government customers only.

Utilizes paramagnetic bead-based purification system for PCR clean up

Manufacturer: Axygen Scientific MAGPCRNM250

 View all options for this product

Catalog No. 14-223-156

  Check Availability
Add to Cart


    For Use With (Application) PCR clean up
    Format 96 and 384-well plate
    No. of Reactions 1920 Preps
    Product Type Kit
    Quantity 25mL

    AxyPrep Mag PCR Normalizer beads have limited binding surface, therefore by limiting the amount of beads added in a given purification reaction, a pre-defined amount DNA can be isolated based on customers need. PCR normalization is accomplished during this purification process so that additional DNA quantification and dilution are not necessary. Time, labor and reagent cost are greatly saved with the AxyPrep Mag unique normalization purification chemistry and thus efficiently streamlining genomics down stream applications. The protocol mainly consists of binding, washing and elution steps, and can be performed directly in the thermal cycling plate and requires no centrifugation or filtration. The process can be automated with walk away solution for high throughput applications.

    • Dual action: Normalization and PCR clean up enables significant reduction in processing and faster turn around times
    • Scalable: Tube, 96-well and 384-well plate formats
    • Manual and automation friendly protocol
    • Compatible with several downstream applications
    • No centrifugation or filtration required

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit