Axygen™ AxyPrep™ Mag PCR Normalizer Kits Available on GSA/VA Contract for Federal Government customers only.

Utilizes paramagnetic bead-based purification system for PCR clean up

Manufacturer: Axygen Scientific MAGPCRNM5

 View more versions of this product

Catalog No. 14-223-154

  Check Availability
Add to Cart

    Specifications

    Format 96 and 384-well plate
    No. of Reactions 96 Preps
    Product Type Kit
    Quantity 5mL,5mL
    Product Name Kit
    Sufficient For 96 Preps
    For Use With (Application) PCR clean up

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit