Learn More
Axygen™ Axyprep™ MAG Plant Genomic DNA Extraction Kit
Magnetic beads based purification system allows rapid and reliable isolation of genomic DNA from a wide variety of plant samples. Up to 96 samples (50 mg fresh tissue per sample, or 15 mg of dried tissue per sample) can be processed in less than an hour.
$183.00 - $1049.00
Specifications
For Use With (Application) | DNA extraction and purification |
---|---|
Product Type | MAG Plant Genomic DNA Extraction Kit |
Catalog Number | Mfr. No. | Includes | No. of Reactions | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | No. of Reactions | Price | Quantity & Availability | ||||
14-223-418
|
Axygen™
MAGPLANTGDNA10 |
MAG-A1 Particles 250μL; PCL Buffer 8 mL; PDB Buffer 7 mL; HSW Buffer 4 mL; MB Elution Buffer 1.5 mL; Protocol Manual 1 | 10 Preps |
Case for $183.00
|
|
||||
14-223-420
|
Axygen™
MAGPLANTGDNAM |
MAG-A1 Particles 10 mL; PCL Buffer 250 mL; PDB Buffer 250 mL; HSW Buffer 100 mL; MB Elution Buffer 60 mL; Protocol Manual 1 | 384 Preps |
Case for $1,049.00
|
|
||||
14-223-419
|
Axygen™
MAGPLANTGDNAS |
MAG-A1 Particles 2.2 mL; PCL Buffer 60 mL; PDB Buffer 60 mL; HSW Buffer 44 mL; MB Elution Buffer 15 mL; Protocol Manual 1 | 96 Preps |
Case for $569.00
|
|
||||
Description
The AxyPrep MAG Plant gDNA Extraction Kit is a magnetic beads-based purification system that allows rapid and reliable isolation of high quality gDNA from a wide variety of plant samples. Up to 96 samples (50 mg fresh tissue per sample or 15 mg of dried tissue per sample) can be processed in less than one hour. The AxyPrep MAG Plant gDNA Extraction Kit efficiently eliminates polysaccharides, phenolic compounds and enzyme inhibitors with no organic extractions. This kit is adaptable for use on major liquid handling workstations for high throughput processing.
- Simplified extraction process
- Non-alcohol-based purification
- Compatible with genomics downstream applications
- High quality, high-yield DNA
- Manual and automation friendly
Specifications
DNA extraction and purification | |
MAG Plant Genomic DNA Extraction Kit |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.