Axygen™ AxyPrep™ MAG Soil, Stool and Water DNA Kit Available on GSA/VA Contract for Federal Government customers only.
One kit for several sample types - Ideal for environmental sample testing
Manufacturer: Axygen Scientific MAGSTLM
 View more versions of this product
Catalog No. 14-223-417
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) DNA extraction and purification
    Includes MAG-A1 Particles: 1.1; PTL Buffer: 2x300mL; Glass Beads: 50g; IR Reagent: 50mL; Binding Enhancer: 3mL; RNase A: 500μL; Pro K Solution: 5mL; Protocol Manual:1
    Product Type AxyPrep Mag Soil, Stool and Water DNA Kit
    Quantity Medium; 384 Preps
    Allows rapid and reliable isolation of high-quality total DNA from fresh and frozen stool samples
    Up to 200 mg stool samples can be processed in less than 60 minutes
    Combines the reversible nucleic acid binding properties of MAG-A1 particles with the efficiency of IR Reagent to eliminate humic acid, polysaccharides, phenolic compounds, and enzyme inhibitors
    Purified DNA is suitable for PCR, restriction digestion, and hybridization techniques
    No organic extractions, which reduces plastic waste and hands-on time allowing multiple samples to be processed in parallel
    • Simplified DNA concentration process: 96 well microplate samples in less than an hour
    • Inhibitor-free DNA purification process
    • Compatible wtih genomics downstream applications
    • Manual and automation friendly
    • PCR and qPCR
    • Sequencing: Sanger and Next Generation
    • Genotyping and SNP detection
    • Restriction digestion
  • Description & Specifications
Related Products
Product Content Feedback