Axygen™ Axyprep™ MAG Viral NA Kit Available on GSA/VA Contract for Federal Government customers only.
Designed for rapid and reliable isolation of DNA from whole blood, serum, plasma, saliva and other bodily fluids
Manufacturer: Axygen Scientific MAGVNAS
 View more versions of this product
Catalog No. 14-223-424
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For rapid and reliable isolation of DNA from whole blood, serum, plasma, saliva and other bodily fluids
    Product Type MAG Viral NA Kit
    Quantity Small; 96 Preps
    High quality DNA is suitable for direct use in most downstream applications such as amplification and enzymatic reactions. The kit can be adapted to most major liquid handling workstations in the market.
    Simplified extraction process
    High quality and yield of nucleic acid purification
    Manual and automation friendly
    Compatible with genomic downstream applications: PCR, Sanger and next generation sequencing, genotyping and SNP detection, and gene expression
  • Description & Specifications
Related Products
Product Content Feedback