Learn More
Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips
Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy
Manufacturer:  Axygen Scientific BT250LRS
 View more versions of this product
Catalog No. 14-222-052
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Robotic Tip
Sterile
Black
Round
10 x 96/Rack
RNase, DNase-free, Non-ANSI
Beckman™ Coulter™ Biomek™ 1000 and 2000 series automation systems
250μL
Clear
Nonfilter, Maxymum Recovery
Polypropylene
96 Tips/Rack
Maxymum Recovery™, non-pyrogenic, uses unique diamond polished molding technology, use no mold-releasing agents, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination, minimize waste of precious samples