Axygen™ Beckman™ Biomek 1000 and 2000 Robotic Tips Available on GSA/VA Contract for Federal Government customers only.

Maxymum Recovery™ reduces sample loss and improves reproducibility and accuracy

Manufacturer: Axygen Scientific BTF20RS

 View more versions of this product

Catalog No. 14-222-059

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Filtered Robotic Tip
    Material Polypropylene
    Packaging 10 x 96/Rack
    Quantity 96 Tips/Rack
    For Use With (Equipment) Beckman™ Coulter™ Biomek™ 1000 and 2000 series automation systems
    Sterility Sterile
    Tip Style Filter
    Shape Round
    Certifications/Compliance RNase, DNase-free, Non-ANSI
    For Use With (Application) Non-pyrogenic, uses unique diamond polished molding technology, use no mold-releasing agents, nucleic acid, nuclease and endotoxin-safe for complete protection against contamination, minimize waste of precious samples and reagents
    Volume (Metric) 20μL
    Color Clear
    Rack Color Gray

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit