Axygen™ Biomek™ FX/NX Aerosol Barrier Filtered Robotic Tips Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Axygen Scientific FXF384RS
 View more versions of this product
Catalog No. 14-222-124
  Check Availability
Add to Cart
    Specifications
    Quantity 384 Tips/Rack
    Sterility Sterile
    Product Type Filtered Robotic Tip
    Material Polypropylene
    For Use With (Equipment) Beckman™ Coulter™ Biomek™ FX, Biomek™ NX and Multimek™ AP-384 head, Dynamic Devices Oasis™ LM 384-Channel-Low Volume, Hamilton™ M
    Packaging Rack
    Color Clear
    Autoclavable Yes
    Tip Style Filter
    Shape Round
    Certifications/Compliance DNase, RNase, pyrogen free
    Volume (Metric) 25μL
    Rack Color Green
  • Description & Specifications
Related Products
Product Content Feedback