Learn More
Axygen™ Biomek™ FX/NX Robotic Tips
Manufacturer:  Axygen Scientific FX255WBRS
 View more versions of this product
Catalog No. 14-222-097
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
  • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
  • Unsurpassed straightness to ensure tip enters the well smoothly, every time
  • 384/1536-well compatible to meet your most demanding applications
Description & Specifications
Specifications
Robotic Tip
Sterile
Blue
Wide Bore
Rack
RNase, DNase-free, ANSI
Beckman™ Coulter™ Biomek™ FX, Biomek™ NX, Biomek™ 3000 and Multimek™ automated liquid handling instruments with AP-96, SPAN-8, MP20, MP200 and AP-3
250μL
Clear
Wide Bore
Polypropylene
384 Tips/Rack
Non-pyrogenic, used for stringent specifications under strict process controls, for pipetting viscous materials, genomic DNA or delicate cell cultures