Axygen™ Biomek™ FX/NX Robotic Tips

Manufacturer: Axygen Scientific FX384LRS

 View more versions of this product

Catalog No. 14-222-101

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Robotic Tip
    Material Polypropylene
    Quantity 384 Tips/Rack
    For Use With (Equipment) Beckman™ Coulter™ Biomek™ FX, Biomek™ NX, Biomek™ 3000 and Multimek™ automated liquid handling instruments with AP-96, SPAN-8, MP20, MP200 and AP-3
    Sterility Sterile
    Tip Style Maxymum Recovery
    Shape Round
    Packaging Rack
    Certifications/Compliance RNase, DNase-free, ANSI
    For Use With (Application) Non-pyrogenic, Maxymum Recovery™, used for stringent specifications under strict process controls
    Volume (Metric) 0.5 to 30μL
    Color Clear
    Rack Color Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit