Axygen™ Biomek™ FX/NX Robotic Tips

Manufacturer: Axygen Scientific FX10RS

 View more versions of this product

Catalog No. 14-222-085

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Robotic Tip
    Volume (Metric) 10μL
    Sterility Sterile
    Color Clear
    Rack Color Violet
    Shape Round
    Material Polypropylene
    Packaging Rack
    Quantity 96 Tips/Rack
    Certifications/Compliance RNase, DNase-free, ANSI
    For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
    For Use With (Equipment) Beckman™ Coulter™ Biomek™ FX, Biomek™ NX, Biomek™ 3000 and Multimek™ automated liquid handling instruments with AP-96, SPAN-8, MP20, MP200 and AP-3

    The Axygen brand range of products includes a comprehensive line of tips for Beckman Coulter automated liquid handling systems. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. The pipette tips are also available with MAXYMum Recovery, a unique low-retention technology that improves tip performance and maximizes the use of your precious samples.

    • Available in racked, racked sterile and with aerosol barrier (filtered)
    • Compatible with Beckman Biomek FX and Beckman Multimek instruments with an AP96 head
    • Also compatible with Span-8 pod when Liquid Level Sensing function is turned off
    • Unsurpassed straightness to ensure tip enters the well smoothly, every time
    • 384/1536-well compatible to meet your most demanding applications
    • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
    • Fine Point improves accuracy and precision at lower sample volumes
    • Maxymum recovery™ tips reduce sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit