Axygen™ Biomek™ FX/NX Robotic Tips Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Axygen Scientific FX50RS

 View more versions of this product

Catalog No. 14-222-111

  Check Availability
Add to Cart

    Specifications

    Sterility Sterile
    Product Type Robotic Tip
    Material Polypropylene
    Quantity 96 Tips/Rack
    For Use With (Equipment) Beckman™ Coulter™ Biomek™ FX, Biomek™ NX, Biomek™ 3000 and Multimek™ automated liquid handling instruments with AP-96, SPAN-8, MP20, MP200 and AP-3
    Shape Round
    Packaging Rack
    Certifications/Compliance RNase, DNase-free, ANSI
    For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
    Volume (Metric) 50μL
    Color Clear
    Rack Color Green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit