Axygen™ Biomek™ FX/NX Robotic Tips Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Axygen Scientific FX153630FPRS
 View more versions of this product
Catalog No. 14-222-087
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    Product Type Robotic Tip
    Material Polypropylene
    Quantity 384 Tips/Rack
    For Use With (Equipment) Beckman™ Coulter™ Biomek™ FX, Biomek™ NX, Biomek™ 3000 and Multimek™ automated liquid handling instruments with AP-96, SPAN-8, MP20, MP200 and AP-3
    Sterility Sterile
    Tip Style Fine-Point
    Packaging Rack
    Certifications/Compliance RNase, DNase-free, ANSI
    For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls, fine point improves accuracy and precision at lower sample volumes
    Volume (Metric) 30μL
    Color Clear
    Rack Color Green
  • Description & Specifications
Related Products
Product Content Feedback