Axygen™ Biomek™ FX/Wellpro™ Robotic Tips
For Beckman FX/Wellpro system
Manufacturer: Axygen Scientific FX384XLLRS
 View more versions of this product
Catalog No. 14-222-105
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Robotic Tip
    Volume (Metric) 50μL
    Sterility Sterile
    Color Clear
    Autoclavable Yes
    Tip Style Extended, Racked
    Material Polypropylene
    Packaging Rack
    Quantity 384 Tips/Rack
    Certifications/Compliance DNase, RNase, pyrogen free
    For Use With (Application) Fine point improves accuracy and precision at lower sample volumes, Maxymum Recovery™ tip reduces sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents
    For Use With (Equipment) Beckman™ Coulter™ Biomek™ FX, Biomek™ NX and Multimek™ AP-384 head, Dynamic Devices Oasis™ LM 384-Channel-Low Volume, Hamilton™ M
    • Tip color: Clear
    • Rack color: Green
    • Volume range: 0.5- 50µL
    • Fine Point improves accuracy and precision at lower sample volumes
    • Extended length - allows pipetting from bottom of 384 deep-well and 384/1536-microplates
    • Maxymum recovery tip reduces sample loss, improves reproducibility and accuracy, minimizes usage of precious samples and reagents
  • Description & Specifications
Related Products
Product Content Feedback