Axygen™ Biomek™ FX/Wellpro™ Robotic Tips Available on GSA/VA Contract for Federal Government customers only.

For Beckman FX/Wellpro system

Manufacturer: Axygen Scientific FX384XLRS

 View more versions of this product

Catalog No. 14-222-107

  Check Availability
Add to Cart

    Specifications

    Sterility Sterile
    Product Type Robotic Tip
    Material Polypropylene
    Quantity 384 Tips/Rack
    For Use With (Equipment) Beckman™ Coulter™ Biomek™ FX, Biomek™ NX and Multimek™ AP-384 head, Dynamic Devices Oasis™ LM 384-Channel-Low Volume, Hamilton™ M
    Autoclavable Yes
    Tip Style Extended, Racked
    Packaging Rack
    Certifications/Compliance DNase, RNase, pyrogen free
    For Use With (Application) Fine point improves accuracy and precision at lower sample volumes
    Volume (Metric) 50μL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit