Axygen Caps for PCR Tubes

8-Strip domed or flat strip caps for Axygen PCR tubes

Manufacturer: Axygen Scientific PCR02CPR

 View more versions of this product

Catalog No. 14-222-268

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Closure Material Polypropylene
    Format Strip
    For Use With (Equipment) 8-strips PCR tubes and 96-well PCR microplates
    Capacity (Metric) 0.2mL
    Type 8-Strip Domed PCR Tube Cap
    Quantity 125 Pack
    Sterile No
    Cap Type Domed
    Certifications/Compliance RNase, DNase, pyrogen-free
    Closure Color Red
    Color Red
    Material Polypropylene

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit