Learn More
Axygen Caps for PCR Tubes
8-Strip domed or flat strip caps for Axygen PCR tubes
Manufacturer:  Axygen Scientific PCR02CPV
 View more versions of this product
Catalog No. 14-222-269
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications
Description
0.2mL with thin walls in various colors.
  • Certified RNase-/DNase-free
  • Nonpyrogenic
  • Nonsterile
  • Manufactured from polypropylene
Description & Specifications
Specifications
8-Strip Domed PCR Tube Cap
RNase, DNase, pyrogen-free
Violet
125 Pack
Polypropylene
8-strips PCR tubes and 96-well PCR microplates
Domed
Violet
Polypropylene
No
Strip
0.2mL