Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Disposable V-Bottom Reagent Reservoirs Available on GSA/VA Contract for Federal Government customers only.

Designed to accommodate all multichannel pipettor

Manufacturer:  Axygen Scientific RESV50

 View more versions of this product

Catalog No. 14-222-433


  Check Availability
Add to cart

Specifications

Description

Four corners and sharp V-bottom shape ensures maximum sample retrieval.

Description & Specifications

Specifications

No
50mL
ANSI, RNase/DNase-free, non-pyrogenic
1.69 oz., 50mL
Yes
1.69 oz.
Polystyrene
100/Cs.
White
Reagent Reservoir
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit