Axygen™ Disposable V-Bottom Reagent Reservoirs Available on GSA/VA Contract for Federal Government customers only.

Designed to accommodate all multichannel pipettor

Manufacturer: Axygen Scientific RES2CV25

 View more versions of this product

Catalog No. 14-222-398

  Check Availability
Add to Cart

    Specifications

    Material Polystyrene
    Color White
    Disposable Yes
    Product Type Reagent Reservoir
    Capacity (Metric) 25mL
    Certifications/Compliance ANSI, RNase/DNase-free, non-pyrogenic
    Packaging Divided; 100/Cs.
    Capacity 0.84 oz., 25mL
    Sterile No
    Capacity (English) 0.84 oz.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit