Learn More
Axygen™ Gel Documentation System Accessory, Optional Filters
For use with Gel Documentation System (GDBL-1000)
Supplier: Axygen™ GDBLRED
Description
Note: For research use only. Not intended for use in diagnostic or therapeutic procedures.Specifications
Red | |
Gel Documentation System (GDBL-1000) | |
676nm | |
Filter |