Learn More
Axygen™ Gel Tips, Round Available on GSA/VA Contract for Federal Government customers only.
Universal fit for research grade pipetters
$92.10 - $99.00
Specifications
Clean Claims DNase, RNase Free
Graduations 2, 5μL
Material PP (Polypropylene)
Shape Round
Tip Style Gel Loading
View More Specs
Products 2
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
14-222-809
View Documents
Axygen™
TGL200RD57R
Non-sterile Case of 400 for $92.10
Add to cart
14-222-810 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Axygen™
TGL200RD57RS
Sterile Case of 400 for $99.00
Add to cart
Description
Description
Axygen™ Gel Loading Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brand of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. Available in either flat or round versions, with different outer diameters for different gel thickness.
  • Designed for removing bands from agarose gel
  • One handed operation
  • Push button gel release
  • Ejector button tip release
  • Avoid cross contamination
  • Tips fit standard 200 µL pipettors
  • These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
  • Nonpyrogenic
  • Nonsterile
  • This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
    Specifications
    Specifications
    DNase, RNase Free
    PP (Polypropylene)
    Gel Loading
    0.5 to 200 μL
    2, 5μL
    Round
    Clear