Learn More
Axygen™ GentleFit™ Dual-Shot Filter Pipet Tips
Soft, leakproof seal provides a universal fit
Manufacturer:  Axygen Scientific TLF1000RS
 View more versions of this product
Catalog No. 14-223-414
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
These filter tips are manufactured from two different resins. The thermoplastic elastomer (TPE) top is easy to seal, provides a soft eject, and is colored-coded by volume. The rigid, polypropylene body ensures straightness, affords glass-like clarity and is non-reactive and graduated.
  • Versatile; dual-shot technology standardizes most research-grade pipetters
  • Available in three sizes: 10, 200 and 1000µL
  • Reduced sealing force
  • Minimal reciprocal ejection force
  • Economical; imposes less wear on pipetters
  • Ergonomic design; soft tip requires less pressure to create an optimum seal
  • Ideal for multichannel pipetters
  • Color-coded top identifies volume
  • Presterilized
  • Certified to be free from RNase, DNase, and human DNA
Specifications
Specifications
Rack
1000μL
Filtered