Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ Impermamat AxyMat™ Sealing Mats for 2mL 96 Well Plates with Square Wells

Chemical resistant mat minimizes evaporation in wide range of temperatures in PCR and storage applications

Manufacturer:  Axygen Scientific AM2MLSQIMP

 View more versions of this product

Catalog No. 14-222-013


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Impermamat Sealing Mat
-176° to 250°F
Sealing Mat
No
96
5
10.0
Yes
-80° to 121°C
Silicone
Natural
2mL 96-well microplates
Square
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit