Axygen™ LightCycler 480 Compatible PCR Plates

384-well plate is designed to fit the Roche LightCycler™ 480

Manufacturer: Axygen Scientific PCR384LC480CNF

 View more versions of this product

Catalog No. 14-223-320

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) RT-PCR
    For Use With (Equipment) For Roche light cycler 480
    Includes N/A
    Certifications/Compliance RNase, DNase-free
    Volume (Metric) Working 3μL
    Color Clear
    Product Type 384-Well PCR Microplate
    Applications RT-PCR

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit