Axygen™ LightCycler 480 Compatible PCR Plates

384-well plate is designed to fit the Roche LightCycler™ 480

Manufacturer: Axygen Scientific PCR384LC480WBC

 View more versions of this product

Catalog No. 14-223-212

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) RT-PCR
    For Use With (Equipment) For Roche light cycler 480
    Includes Sealing film, barcodes
    Certifications/Compliance RNase, DNase-free
    Volume (Metric) Working 3μL
    Color White
    Product Type 384-Well PCR Microplate
    Applications RT-PCR

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit