Learn More
Axygen™ Low Profile 8-Strip PCR Tubes
Axygen™ brand low profile 8-strip PCR tubes are ideal for low sample volume reactions.
Manufacturer:  Axygen Scientific PCR0108LPW
 View more versions of this product
Catalog No. 14-223-507
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Using highly polished and precise molds, our tubes ensure precise thermal transfer and a leak-proof seal with a choice of strip caps.
  • Low profile design results in faster and more efficient PCR reactions
  • Ideal for 0.1mL thermal cyclers
  • Available in white or clear
  • Real time optimized clear PCR 8-strip caps, domed 8-strip caps and 8-strip frosted flat caps
  • Certified DNase-/RNase-free and nonpyrogenic
  • Recommended volume range = 5µL to 125µL
Specifications
Specifications
Non-sterile
RNase, DNase and non-pyrogenic free
Polypropylene
125 Pack
0.1mL thermal cycler for low sample volume reactions
Thin Wall 8-Strip PCR Tube
White
0.1mL
Strip
Conical